2-[4-(1-Methyl-1-phenylethyl)phenoxy]acetohydrazide

Supplier: Matrix Scientific

029738-500MG
101828-758EA 109.25 USD
101828-758
2-[4-(1-Methyl-1-phenylethyl)phenoxy]acetohydrazide
2-[4-(1-Methyl-1-phenylethyl)phenoxy]acetohydrazide

Hazard: Irritant

Formula: C₁₇H₂₀N₂O₂
MDL Number: MFCD02351180
CAS Number: 70757-64-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR