2-(2,3-Dihydro-1H-inden-5-yloxy)acetohydrazide
Supplier: Matrix Scientific
029735-500MG
101828-752EA
146.15
USD
101828-752
2-(2,3-Dihydro-1H-inden-5-yloxy)acetohydrazide
2-(2,3-Dihydro-1H-inden-5-yloxy)acetohydrazide
Hazard: Irritant
Formula:
C₁₁H₁₄N₂O₂ |
MDL Number:
MFCD03423117 CAS Number: 667437-07-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...