2-(2-Nitrophenoxy)acetohydrazide

Supplier: Matrix Scientific

029733-500MG
101828-748EA 146.15 USD
101828-748
2-(2-Nitrophenoxy)acetohydrazide
2-(2-Nitrophenoxy)acetohydrazide

Hazard: Irritant

Formula: C₈H₉N₃O₄
MDL Number: MFCD00806916
CAS Number: 77729-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR