2-(4-tert-Butylphenoxy)acetohydrazide

Supplier: Matrix Scientific

029730-500MG
101828-742EA 109.25 USD
101828-742
2-(4-tert-Butylphenoxy)acetohydrazide
2-(4-tert-Butylphenoxy)acetohydrazide

Hazard: Irritant

Formula: C₁₂H₁₈N₂O₂
MDL Number: MFCD01536446
CAS Number: 75843-50-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR