2-(4-Propylphenyl)quinoline-4-carbohydrazide

Supplier: Matrix Scientific

029729-500MG
101828-740EA 290.77 USD
101828-740
2-(4-Propylphenyl)quinoline-4-carbohydrazide
2-(4-Propylphenyl)quinoline-4-carbohydrazide

Hazard: Irritant

Formula: C₁₉H₁₉N₃O
MDL Number: MFCD02244950
CAS Number: 524934-10-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR