2-(2,5-Dichlorophenoxy)acetohydrazide

Supplier: Matrix Scientific

029728-500MG
101828-738EA 146.15 USD
101828-738
2-(2,5-Dichlorophenoxy)acetohydrazide
2-(2,5-Dichlorophenoxy)acetohydrazide

Hazard: Irritant

Formula: C₈H₈Cl₂N₂O₂
MDL Number: MFCD00980669
CAS Number: 32022-40-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR