2-(3,4-Dimethylphenoxy)acetohydrazide

Supplier: Matrix Scientific

029727-500MG
101828-736EA 146.15 USD
101828-736
2-(3,4-Dimethylphenoxy)acetohydrazide
2-(3,4-Dimethylphenoxy)acetohydrazide

Hazard: Irritant

Formula: C₁₀H₁₄N₂O₂
MDL Number: MFCD00815926
CAS Number: 125298-97-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR