2-(3-Methyl-4-nitrophenoxy)acetohydrazide

Supplier: Matrix Scientific

029724-500MG
101828-730EA 146.15 USD
101828-730
2-(3-Methyl-4-nitrophenoxy)acetohydrazide
2-(3-Methyl-4-nitrophenoxy)acetohydrazide

Hazard: Irritant

Formula: C₉H₁₁N₃O₄
MDL Number: MFCD03423115
CAS Number: 588679-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR