2-(3-Chloro-4-fluorophenoxy)acetohydrazide

Supplier: Matrix Scientific

029721-500MG
101828-724EA 109.25 USD
101828-724
2-(3-Chloro-4-fluorophenoxy)acetohydrazide
2-(3-Chloro-4-fluorophenoxy)acetohydrazide

Hazard: Irritant

Formula: C₈H₈ClFN₂O₂
MDL Number: MFCD03423112
CAS Number: 774575-30-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR