5-[(3,5-Dimethylphenoxy)methyl]-2-furohydrazide

Supplier: Matrix Scientific

029718-500MG
101828-718EA 217.35 USD
101828-718
5-[(3,5-Dimethylphenoxy)methyl]-2-furohydrazide
5-[(3,5-Dimethylphenoxy)methyl]-2-furohydrazide

Hazard: Irritant

Formula: C₁₄H₁₆N₂O₃
MDL Number: MFCD02253813
CAS Number: 438221-61-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR