5-[(4-Bromo-1H-pyrazol-1-yl)methyl]-2-furohydrazide

Supplier: Matrix Scientific

029712-500MG
101828-706EA 290.77 USD
101828-706
5-[(4-Bromo-1H-pyrazol-1-yl)methyl]-2-furohydrazide
5-[(4-Bromo-1H-pyrazol-1-yl)methyl]-2-furohydrazide

Hazard: Irritant

Formula: C₉H₉BrN₄O₂
MDL Number: MFCD02055805
CAS Number: 402729-65-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR