5-[(4-Chloro-3,5-dimethyl-1H-pyrazol-1-yl)methyl]-2-furohydrazide

Supplier: Matrix Scientific

029711-500MG
101828-704EA 217.35 USD
101828-704
5-[(4-Chloro-3,5-dimethyl-1H-pyrazol-1-yl)methyl]-2-furohydrazide
5-[(4-Chloro-3,5-dimethyl-1H-pyrazol-1-yl)methyl]-2-furohydrazide

Hazard: Irritant

Formula: C₁₁H₁₃ClN₄O₂
MDL Number: MFCD02253817
CAS Number: 386705-59-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR