8-Chloro-2-(4-ethylphenyl)quinoline-4-carbohydrazide

Supplier: Matrix Scientific

029710-500MG
101828-702EA 290.77 USD
101828-702
8-Chloro-2-(4-ethylphenyl)quinoline-4-carbohydrazide
8-Chloro-2-(4-ethylphenyl)quinoline-4-carbohydrazide

Hazard: Irritant

Formula: C₁₈H₁₆ClN₃O
MDL Number: MFCD03422998
CAS Number: 588711-80-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR