8-Chloro-2-phenylquinoline-4-carbohydrazide

Supplier: Matrix Scientific

029709-500MG
101828-700EA 217.35 USD
101828-700
8-Chloro-2-phenylquinoline-4-carbohydrazide
8-Chloro-2-phenylquinoline-4-carbohydrazide

Hazard: Irritant

Formula: C₁₆H₁₂ClN₃O
MDL Number: MFCD03422952
CAS Number: 854867-67-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR