2-(2,5-Dimethoxyphenyl)quinoline-4-carbohydrazide

Supplier: Matrix Scientific

029708-500MG
101828-698EA 290.77 USD
101828-698
2-(2,5-Dimethoxyphenyl)quinoline-4-carbohydrazide
2-(2,5-Dimethoxyphenyl)quinoline-4-carbohydrazide

Hazard: Irritant

Formula: C₁₈H₁₇N₃O₃
MDL Number: MFCD03422939
CAS Number: 438214-25-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR