3-Methylthiophene-2-carbohydrazide

Supplier: Matrix Scientific

029707-500MG
101828-696EA 193.85 USD
101828-696
3-Methylthiophene-2-carbohydrazide
3-Methylthiophene-2-carbohydrazide

Hazard: Irritant

Formula: C₆H₈N₂OS
MDL Number: MFCD01993658
CAS Number: 350997-56-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR