3,4-Dichloro-1-benzothiophene-2-carbohydrazide

Supplier: Matrix Scientific

Synonyms: OGG1 inhibitor O83,4-Dichloro-benzo[b]thiophene-2-carboxylic acid hydrazideOGG1-IN-083,4-Dichlorobenzo[b]thiophene-2-carbohydrazide

029706-500MG
101828-694EA 290.77 USD
101828-694
3,4-Dichloro-1-benzothiophene-2-carbohydrazide
3,4-Dichloro-1-benzothiophene-2-carbohydrazide

Hazard: Irritant

Formula: C₉H₆Cl₂N₂OS
MW: 261.13 g/mol
MDL Number: MFCD01993639
CAS Number: 350997-39-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR