3-Chloro-6-ethyl-1-benzothiophene-2-carbohydrazide

Supplier: Matrix Scientific

029705-500MG
101828-692EA 217.35 USD
101828-692
3-Chloro-6-ethyl-1-benzothiophene-2-carbohydrazide
3-Chloro-6-ethyl-1-benzothiophene-2-carbohydrazide

Hazard: Irritant

Formula: C₁₁H₁₁ClN₂OS
MDL Number: MFCD01924130
CAS Number: 351000-82-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR