4-(Hydrazinocarbonyl)benzamide

Supplier: Matrix Scientific

029700-500MG
101828-682EA 173.85 USD
101828-682
4-(Hydrazinocarbonyl)benzamide
4-(Hydrazinocarbonyl)benzamide

Hazard: Irritant

Formula: C₈H₉N₃O₂
MDL Number: MFCD03422938
CAS Number: 22590-92-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR