Palmitic acid hydrazide

Supplier: Matrix Scientific

Synonyms: Palmitohydrazide

029699-500MG
101828-680EA 146.15 USD
101828-680
Palmitic acid hydrazide
Palmitic acid hydrazide

Hazard: Irritant

Formula: C₁₆H₃₄N₂O
MW: 270.46 g/mol
MDL Number: MFCD00066357
CAS Number: 2619-88-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR