3-(3,4-Dihydroxyphenyl)propanohydrazide

Supplier: Matrix Scientific

029697-500MG
101828-676EA 146.15 USD
101828-676
3-(3,4-Dihydroxyphenyl)propanohydrazide
3-(3,4-Dihydroxyphenyl)propanohydrazide

Hazard: Irritant

Formula: C₉H₁₂N₂O₃
MDL Number: MFCD03014983
CAS Number: 401642-48-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR