2-Cyclohexylacetohydrazide

Supplier: Matrix Scientific

029696-500MG
101828-674EA 153.85 USD
101828-674
2-Cyclohexylacetohydrazide
2-Cyclohexylacetohydrazide

Hazard: Irritant

Formula: C₈H₁₆N₂O
MDL Number: MFCD02244577
CAS Number: 27563-60-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR