5-Chloro-2-hydroxybenzhydrazide
Supplier: Matrix Scientific
029694-500MG
101828-670EA
109.25
USD
101828-670
5-Chloro-2-hydroxybenzhydrazide
5-Chloro-2-hydroxybenzhydrazide
Hazard: Irritant
Formula:
C₇H₇ClN₂O₂ |
MDL Number:
MFCD01001347 CAS Number: 5022-48-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...