1-Benzyl-2,3-dimethyl-1H-indole-5-carbohydrazide

Supplier: Matrix Scientific

029692-500MG
101828-666EA 144.9 USD
101828-666
1-Benzyl-2,3-dimethyl-1H-indole-5-carbohydrazide
1-Benzyl-2,3-dimethyl-1H-indole-5-carbohydrazide

Hazard: Irritant

Formula: C₁₈H₁₉N₃O
MDL Number: MFCD01922190
CAS Number: 350990-21-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR