2-Thien-2-ylacetohydrazide

Supplier: Matrix Scientific

029690-500MG
101828-662EA 109.25 USD
101828-662
2-Thien-2-ylacetohydrazide
2-Thien-2-ylacetohydrazide

Hazard: Irritant

Formula: C₆H₈N₂OS
MDL Number: MFCD00985059
CAS Number: 39978-18-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR