2-(2,6-Dichlorophenyl)acetohydrazide
Supplier: Matrix Scientific
029689-500MG
101828-660EA
146.15
USD
101828-660
2-(2,6-Dichlorophenyl)acetohydrazide
2-(2,6-Dichlorophenyl)acetohydrazide
Hazard: Irritant
Formula:
C₈H₈Cl₂N₂O |
MDL Number:
MFCD01923147 CAS Number: 129564-34-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...