2-(3,4-Dichlorophenyl)acetohydrazide

Supplier: Matrix Scientific

Synonyms: 3,4-Dichlorophenylacetic acid hydrazide

029688-500MG
101828-658EA 146.15 USD
101828-658
2-(3,4-Dichlorophenyl)acetohydrazide
2-(3,4-Dichlorophenyl)acetohydrazide

Hazard: Irritant

Formula: C₈H₈Cl₂N₂O
MW: 219.07 g/mol
MDL Number: MFCD00833407
CAS Number: 129564-33-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR