2-Hydroxy-1-naphthohydrazide

Supplier: Matrix Scientific

029681-500MG
101828-644EA 146.15 USD
101828-644
2-Hydroxy-1-naphthohydrazide
2-Hydroxy-1-naphthohydrazide

Hazard: Irritant

Formula: C₁₁H₁₀N₂O₂
MDL Number: MFCD03422935
CAS Number: 7248-26-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR