2-Methyl-3-furohydrazide

Supplier: Matrix Scientific

Synonyms: 2-Methyl-3-furoic hydrazide

029676-500MG
101828-634EA 146.15 USD
101828-634
2-Methyl-3-furohydrazide
2-Methyl-3-furohydrazide

Hazard: Irritant

Formula: C₆H₈N₂O₂
MW: 140.14 g/mol
MDL Number: MFCD01330023
CAS Number: 315672-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR