4-tert-Butylcyclohexanecarbohydrazide

Supplier: Matrix Scientific

029675-500MG
101828-632EA 165.6 USD
101828-632
4-tert-Butylcyclohexanecarbohydrazide
4-tert-Butylcyclohexanecarbohydrazide

Hazard: Irritant

Formula: C₁₁H₂₂N₂O
MDL Number: MFCD02055941
CAS Number: 438531-50-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR