9H-Xanthene-9-carbohydrazide

Supplier: Matrix Scientific

029674-500MG
101828-630EA 146.15 USD
101828-630
9H-Xanthene-9-carbohydrazide
9H-Xanthene-9-carbohydrazide

Hazard: Irritant

Formula: C₁₄H₁₂N₂O₂
MDL Number: MFCD00100677
CAS Number: 1604-08-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR