2-(4-tert-Butylphenyl)cyclopropanecarbohydrazide

Supplier: Matrix Scientific

029672-500MG
101828-626EA 217.35 USD
101828-626
2-(4-tert-Butylphenyl)cyclopropanecarbohydrazide
2-(4-tert-Butylphenyl)cyclopropanecarbohydrazide

Hazard: Irritant

Formula: C₁₄H₂₀N₂O
MDL Number: MFCD01919592
CAS Number: 438616-66-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR