Cyclopentanecarbohydrazide

Supplier: Matrix Scientific

029669-500MG
101828-620EA 109.25 USD
101828-620
Cyclopentanecarbohydrazide
Cyclopentanecarbohydrazide

Hazard: Irritant

Formula: C₆H₁₂N₂O
Melting Pt: 117…118 °C
MDL Number: MFCD03002174
CAS Number: 3400-07-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR