2,2-Dichloro-1-methylcyclopropanecarbohydrazide

Supplier: Matrix Scientific

029667-500MG
101828-616EA 193.85 USD
101828-616
2,2-Dichloro-1-methylcyclopropanecarbohydrazide
2,2-Dichloro-1-methylcyclopropanecarbohydrazide

Hazard: Irritant

Formula: C₅H₈Cl₂N₂O
MDL Number: MFCD01630709
CAS Number: 351982-73-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR