3,3-Diphenylpropanohydrazide

Supplier: Matrix Scientific

029665-500MG
101828-612EA 146.15 USD
101828-612
3,3-Diphenylpropanohydrazide
3,3-Diphenylpropanohydrazide

Hazard: Irritant

Formula: C₁₅H₁₆N₂O
MDL Number: MFCD01722175
CAS Number: 58973-41-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR