2-Ethylbutanohydrazide

Supplier: Matrix Scientific

029664-500MG
101828-610EA 146.15 USD
101828-610
2-Ethylbutanohydrazide
2-Ethylbutanohydrazide

Hazard: Irritant

Formula: C₆H₁₄N₂O
MDL Number: MFCD03422931
CAS Number: 88141-09-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR