Valeric acid hydrazide
Synonyms:
Pentanehydrazide
029662-500MG
101828-606EA
102.86
USD
101828-606
Valeric acid hydrazide
Valeric acid hydrazide
Hazard: Irritant
Formula:
C₅H₁₂N₂O MW: 116.16 g/mol |
MDL Number:
MFCD00025137 CAS Number: 38291-82-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...