2-Phenyl-2-(phenylthio)acetohydrazide

Supplier: Matrix Scientific

029659-500MG
101828-600EA 146.15 USD
101828-600
2-Phenyl-2-(phenylthio)acetohydrazide
2-Phenyl-2-(phenylthio)acetohydrazide

Hazard: Irritant

Formula: C₁₄H₁₄N₂OS
MDL Number: MFCD03422929
CAS Number: 32121-53-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR