2-Phenylbutanohydrazide

Supplier: Matrix Scientific

029658-500MG
101828-598EA 146.15 USD
101828-598
2-Phenylbutanohydrazide
2-Phenylbutanohydrazide

Hazard: Irritant

Formula: C₁₀H₁₄N₂O
MDL Number: MFCD00119417
CAS Number: 3538-74-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR