3-Isobutoxybenzohydrazide

Supplier: Matrix Scientific

029657-500MG
101828-596EA 109.25 USD
101828-596
3-Isobutoxybenzohydrazide
3-Isobutoxybenzohydrazide

Hazard: Irritant

Formula: C₁₁H₁₆N₂O₂
MDL Number: MFCD01993663
CAS Number: 350997-59-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR