4-Butoxybenzohydrazide

Supplier: Matrix Scientific

029656-500MG
101828-594EA 97.75 USD
101828-594
4-Butoxybenzohydrazide
4-Butoxybenzohydrazide

Hazard: Irritant

Formula: C₁₁H₁₆N₂O₂
MDL Number: MFCD00452662
CAS Number: 64328-61-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR