3-Butoxybenzohydrazide

Supplier: Matrix Scientific

029655-500MG
101828-592EA 146.15 USD
101828-592
3-Butoxybenzohydrazide
3-Butoxybenzohydrazide

Hazard: Irritant

Formula: C₁₁H₁₆N₂O₂
MDL Number: MFCD01993661
CAS Number: 350997-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR