N-[4-(Acetylamino)phenyl]-2-chloroacetamide

Supplier: Matrix Scientific

029650-500MG
101828-582EA 109.25 USD
101828-582
N-[4-(Acetylamino)phenyl]-2-chloroacetamide
N-[4-(Acetylamino)phenyl]-2-chloroacetamide

Hazard: Irritant

Formula: C₁₀H₁₁ClN₂O₂
Melting Pt: 265…270 °C (dec.)
MDL Number: MFCD00186762
CAS Number: 2653-10-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR