N-(4-Bromo-2-methylphenyl)-2-chloroacetamide
Supplier: Matrix Scientific
029649-500MG
101828-580EA
109.25
USD
101828-580
N-(4-Bromo-2-methylphenyl)-2-chloroacetamide
N-(4-Bromo-2-methylphenyl)-2-chloroacetamide
Hazard: Irritant
Formula:
C₉H₉BrClNO |
MDL Number:
MFCD00157388 CAS Number: 96686-51-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...