2-Chloro-N-{4-[(dimethylamino)sulfonyl]-phenyl}acetamide

Supplier: Matrix Scientific

029645-500MG
101828-572EA 217.35 USD
101828-572
2-Chloro-N-{4-[(dimethylamino)sulfonyl]-phenyl}acetamide
2-Chloro-N-{4-[(dimethylamino)sulfonyl]-phenyl}acetamide

Hazard: Irritant

Formula: C₁₀H₁₃ClN₂O₃S
MDL Number: MFCD01671425
CAS Number: 23280-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR