Methyl-3-[(chloroacetyl)amino]thiophene-2-carboxylate

Supplier: Matrix Scientific

029644-500MG
101828-570EA 290.77 USD
101828-570
Methyl-3-[(chloroacetyl)amino]thiophene-2-carboxylate
Methyl-3-[(chloroacetyl)amino]thiophene-2-carboxylate

Hazard: Irritant

Formula: C₈H₈ClNO₃S
MDL Number: MFCD00120696
CAS Number: 146381-88-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR