2-Chloro-N-(5-phenyl-1,3,4-thiadiazol-2-yl)acetamide

Supplier: Matrix Scientific

029642-500MG
101828-566EA 290.77 USD
101828-566
2-Chloro-N-(5-phenyl-1,3,4-thiadiazol-2-yl)acetamide
2-Chloro-N-(5-phenyl-1,3,4-thiadiazol-2-yl)acetamide

Hazard: Irritant

Formula: C₁₀H₈ClN₃OS
MW: 253.71 g/mol
MDL Number: MFCD00500598
CAS Number: 20460-58-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR