2-Chloro-N-[5-(cyclopentylmethyl)-1,3,4-thiadiazol-2-yl]acetamide

Supplier: Matrix Scientific

029638-500MG
101828-558EA 290.77 USD
101828-558
2-Chloro-N-[5-(cyclopentylmethyl)-1,3,4-thiadiazol-2-yl]acetamide
2-Chloro-N-[5-(cyclopentylmethyl)-1,3,4-thiadiazol-2-yl]acetamide

Hazard: Irritant

Formula: C₁₀H₁₄ClN₃OS
MDL Number: MFCD08593621
CAS Number: 908528-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR