2-Chloro-N-[5-(4-methoxyphenyl)-1,3,4-thiadiazol-2-yl]acetamide

Supplier: Matrix Scientific

029637-500MG
101828-556EA 290.77 USD
101828-556
2-Chloro-N-[5-(4-methoxyphenyl)-1,3,4-thiadiazol-2-yl]acetamide
2-Chloro-N-[5-(4-methoxyphenyl)-1,3,4-thiadiazol-2-yl]acetamide

Hazard: Irritant

Formula: C₁₁H₁₀ClN₃O₂S
MDL Number: MFCD00500501
CAS Number: 18199-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR