2-Chloro-N-[4-(morpholin-4-ylsulfonyl)phenyl]acetamide

Supplier: Matrix Scientific

029636-500MG
101828-554EA 290.77 USD
101828-554
2-Chloro-N-[4-(morpholin-4-ylsulfonyl)phenyl]acetamide
2-Chloro-N-[4-(morpholin-4-ylsulfonyl)phenyl]acetamide

Hazard: Irritant

Formula: C₁₂H₁₅ClN₂O₄S
MW: 318.78 g/mol
MDL Number: MFCD02580955
CAS Number: 35959-60-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR